3-Chloro-5-(trifluoromethyl)picolinimidohydrazide

Lieferant: Apollo Scientific
Achtung

PC28272-1G PC28272-250MG
APOSPC28272-1GEA 473 EUR
APOSPC28272-1G APOSPC28272-250MG
3-Chloro-5-(trifluoromethyl)picolinimidohydrazide
3-Chloro-5-(trifluoromethyl)picolinimidohydrazide
Formel: C₇H₆ClF₃N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD31812111
CAS-Nummer: 2379918-35-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR