8-Chloro-3-(chlorodifluoromethyl)-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine

Lieferant: Apollo Scientific
Achtung

PC28276-1G PC28276-250MG
APOSPC28276-1GEA 1360 EUR
APOSPC28276-1G APOSPC28276-250MG
8-Chloro-3-(chlorodifluoromethyl)-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
8-Chloro-3-(chlorodifluoromethyl)-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Formel: C₈H₂Cl₂F₅N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02187042
CAS-Nummer: 478248-86-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR