1-(8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridin-3-yl)-2,2,2-trifluoroethanone

Lieferant: Apollo Scientific
Achtung

PC28278-1G PC28278-250MG PC28278-5G
APOSPC28278-1GEA 432 EUR
APOSPC28278-1G APOSPC28278-250MG APOSPC28278-5G
1-(8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridin-3-yl)-2,2,2-trifluoroethanone
1-(8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridin-3-yl)-2,2,2-trifluoroethanone
Formel: C₁₀H₃ClF₆N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD12406877
CAS-Nummer: 1823182-59-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR