2-Chloro-4-(trifluoromethyl)thiazole-5-carbonitrile

Lieferant: Apollo Scientific
Achtung

PC28295-100MG PC28295-250MG PC28295-1G
APOSPC28295-100MGEA 0 EUR
APOSPC28295-100MG APOSPC28295-250MG APOSPC28295-1G
2-Chloro-4-(trifluoromethyl)thiazole-5-carbonitrile
2-Chloro-4-(trifluoromethyl)thiazole-5-carbonitrile
Formel: C₅ClF₃N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD08669458
CAS-Nummer: 151729-41-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR