3-(Ethylthio)-5-(trifluoromethyl)picolinonitrile

Lieferant: Apollo Scientific
Achtung

PC28297-1G PC28297-250MG
APOSPC28297-1GEA 68.3 EUR
APOSPC28297-1G APOSPC28297-250MG
3-(Ethylthio)-5-(trifluoromethyl)picolinonitrile
3-(Ethylthio)-5-(trifluoromethyl)picolinonitrile
Formel: C₉H₇F₃N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD31812104
CAS-Nummer: 1421952-04-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR