6-Chloro-2-(trifluoromethyl)imidazo[1,2-b]pyridazine

Lieferant: Apollo Scientific

PC28302-1G PC28302-250MG
APOSPC28302-1GEA 302 EUR
APOSPC28302-1G APOSPC28302-250MG
6-Chloro-2-(trifluoromethyl)imidazo[1,2-b]pyridazine
6-Chloro-2-(trifluoromethyl)imidazo[1,2-b]pyridazine
Formel: C₇H₃ClF₃N₃
MDL: MFCD07778347
CAS-Nummer: 109113-97-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR