4-Chloro-7-(trifluoromethoxy)quinoline

Lieferant: Apollo Scientific

PC28306-1G
APOSPC28306-1GEA 197 EUR
APOSPC28306-1G
4-Chloro-7-(trifluoromethoxy)quinoline
4-Chloro-7-(trifluoromethoxy)quinoline
Formel: C₁₀H₅ClF₃NO
Lagertemperatur: Kühlschrank
MDL: MFCD08063185
CAS-Nummer: 40516-31-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR