Perfluoro-3,6-dioxa-4-methyl-7-octenesulphonic acid 95%

Lieferant: Apollo Scientific

PC28310-250MG
APOSPC28310-250MGEA 475 EUR
APOSPC28310-250MG
Perfluoro-3,6-dioxa-4-methyl-7-octenesulphonic acid 95%
Perfluoro-3,6-dioxa-4-methyl-7-octenesulphonic acid
Formel: C₇HF₁₃O₅S
Lagertemperatur: Raumtemperatur
MDL: MFCD32066855
CAS-Nummer: 29311-67-9
UN: 2586
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR