4-Fluorobenzylmagnesium chloride 0,25 M in THF
Lieferant: Apollo Scientific
|
Gefahr
|
PC2833-100ML
APOSPC2833-100MLEA
763
EUR
APOSPC2833-100ML
4-Fluorobenzylmagnesium chloride 0,25 M in THF
4-Fluorobenzylmagnesium chloride
Formel:
C₇H₆ClFMg Molecular Weight: 168,878 g/mol Siedepunkt: 65 °C Dichte: 0,948 g/cm³ (25 °C) Flammpunkt: <–17 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01319898 CAS-Nummer: 1643-73-8 UN: 2924 ADR: 3+8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...