(±)-3,5-Difluormandelsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(±)-3,5-Difluorphenylglykolsäure, (±)-2-(3,5-Difluorphenyl)-2-hydroxyethansäure, (±)-2-(3,5-Difluorphenyl)-2-hydroxyessigsäure
PC2842G-5G
APOSPC2842G-5GEA
66.7
EUR
APOSPC2842G-5G
(±)-3,5-Difluormandelsäure
(±)-3,5-Difluormandelsäure
Formel:
C₈H₆F₂O₃ Molecular Weight: 188.13 g/mol Schmelzpunkt: 135…139 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061300 CAS-Nummer: 132741-31-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...