Hexafluoro-2-methylisopropyl acrylate 98%

Lieferant: Apollo Scientific
Achtung

PC2843-1G PC2843-5G
APOSPC2843-1GEA 105 EUR
APOSPC2843-1G APOSPC2843-5G
Hexafluoro-2-methylisopropyl acrylate 98%
Hexafluoro-2-methylisopropyl acrylate
Formel: C₇H₆F₆O₂
Siedepunkt: 60…61 °C/190mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD03094206
CAS-Nummer: 53038-41-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR