2-(Difluormethoxy)benzonitril 95%

Lieferant: Apollo Scientific

PC2847M-5G
APOSPC2847M-5GEA 66.7 EUR
APOSPC2847M-5G
2-(Difluormethoxy)benzonitril 95%
2-(Difluormethoxy)benzonitril
Formel: C₈H₅F₂NO
Molecular Weight: 169.1282 g/mol
Siedepunkt: 118…120 °C/15mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00221469
CAS-Nummer: 56935-78-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR