4-(Difluormethoxy)benzonitril 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC2847O-5G
PC2847O-25G
PC2847O-50G
APOSPC2847O-5GEA
0
EUR
APOSPC2847O-5G
APOSPC2847O-25G
APOSPC2847O-50G
4-(Difluormethoxy)benzonitril 98%
4-(Difluormethoxy)benzonitril
Formel:
C₈H₅F₂NO Schmelzpunkt: 40…42 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00085006 CAS-Nummer: 90446-25-6 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...