α-Brom-4-(difluormethoxy)toluol 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC2847Q-1G
PC2847Q-25G
PC2847Q-5G
APOSPC2847Q-1GEA
0
EUR
APOSPC2847Q-1G
APOSPC2847Q-25G
APOSPC2847Q-5G
α-Brom-4-(difluormethoxy)toluol 98%
α-Brom-4-(difluormethoxy)toluol
Formel:
C₈H₇BrF₂O Siedepunkt: 64…66 °C/0,3mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085007 CAS-Nummer: 3447-53-8 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...