1-(Difluormethoxy)-2-nitrobenzol 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC2847RS-100G
PC2847RS-25G
PC2847RS-5G
APOSPC2847RS-100GEA
0
EUR
APOSPC2847RS-100G
APOSPC2847RS-25G
APOSPC2847RS-5G
1-(Difluormethoxy)-2-nitrobenzol 97%
1-(Difluormethoxy)-2-nitrobenzol
Formel:
C₇H₅F₂NO₃ Siedepunkt: 73…75 °C/0,3mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00221470 CAS-Nummer: 22225-77-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...