2-Brom-6-(trifluormethyl)benzonitril 95%

Lieferant: Apollo Scientific
Achtung

PC2854-5G PC2854-1G
APOSPC2854-5GEA 404 EUR
APOSPC2854-5G APOSPC2854-1G
2-Brom-6-(trifluormethyl)benzonitril 95%
2-Brom-6-(trifluormethyl)benzonitril
Formel: C₈H₃BrF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD08741382
CAS-Nummer: 914637-35-7
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR