Bis[2-(fluorosulphonyl)tetrafluoroethyl]ether 98%

Lieferant: Apollo Scientific
Gefahr

PC2855-1G
APOSPC2855-1GEA 0 EUR
APOSPC2855-1G
Bis[2-(fluorosulphonyl)tetrafluoroethyl]ether 98%
Bis[2-(fluorosulphonyl)tetrafluoroethyl]ether
Formel: C₄F₁₀O₅S₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03094207
CAS-Nummer: 146829-79-0
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR