2-Brom-6-(trifluormethyl)anilin 98%

Lieferant: Apollo Scientific
Achtung

PC2857-1G PC2857-5G PC2857-10G
APOSPC2857-1GEA 0 EUR
APOSPC2857-1G APOSPC2857-5G APOSPC2857-10G
2-Brom-6-(trifluormethyl)anilin 98%
2-Brom-6-(trifluormethyl)anilin
Formel: C₇H₅BrF₃N
Lagertemperatur: Kühlschrank
MDL: MFCD08741383
CAS-Nummer: 58458-13-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR