2,5-Difluorphenylhydraziniumchlorid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2,5-Difluorphenyl)hydrazin Hydrochlorid, 2,5-Difluorphenylhydrazin Hydrochlorid, 2,5-Difluorphenylhydrazinhydrochlorid

PC2874X-10G PC2874X-5G
APOSPC2874X-10GEA 58 EUR
APOSPC2874X-10G APOSPC2874X-5G
2,5-Difluorphenylhydraziniumchlorid 97%
2,5-Difluorphenylhydraziniumchlorid
Formel: C₆H₇ClF₂N₂
Molecular Weight: 180.58 g/mol
Schmelzpunkt: 210…211 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013385
CAS-Nummer: 175135-73-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR