4-(2,4-Difluorphenyl)-3-thiosemicarbazid 97%

Lieferant: Apollo Scientific

PC2875W-1G
APOSPC2875W-1GEA 72.7 EUR
APOSPC2875W-1G
4-(2,4-Difluorphenyl)-3-thiosemicarbazid 97%
4-(2,4-Difluorphenyl)-3-thiosemicarbazid
Formel: C₇H₇F₂N₃S
Schmelzpunkt: 172…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00060563
CAS-Nummer: 206559-58-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR