2',3'-Difluoropropiophenone 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2,3-Difluorophenyl)propan-1-one

PC2879-10G PC2879-1G PC2879-5G
APOSPC2879-10GEA 128 EUR
APOSPC2879-10G APOSPC2879-1G APOSPC2879-5G
2',3'-Difluoropropiophenone 97%
2',3'-Difluoropropiophenone
Formel: C₉H₈F₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00061141
CAS-Nummer: 236754-63-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR