3',4'-Difluorpropiophenon 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3,4-Difluorophenyl)propan-1-one

PC2883-1G
APOSPC2883-1GEA 21.9 EUR
APOSPC2883-1G
3',4'-Difluorpropiophenon 97%
3',4'-Difluorpropiophenon
Formel: C₉H₈F₂O
Molecular Weight: 170,16 g/mol
Siedepunkt: 64…65 °C/1,8mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00015505
CAS-Nummer: 23384-72-7
EINECS: 245-627-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR