Nonafluoro-tert-butyl isobutyrate
Lieferant: Apollo Scientific
|
Achtung
|
PC2892-1G
APOSPC2892-1GEA
364
EUR
APOSPC2892-1G
Nonafluoro-tert-butyl isobutyrate
Nonafluoro-tert-butyl isobutyrate
Formel:
C₈H₇F₉O₂ Molecular Weight: 306,126 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08741387 CAS-Nummer: 914637-43-7 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...