Methyl-4-hydroxy-3-(trifluoromethyl)benzoate 98+%

Lieferant: Apollo Scientific

Synonyme: Methyl 4-hydroxy-3-(trifluoromethyl)benzoate

PC2901-5G
APOSPC2901-5GEA 115 EUR
APOSPC2901-5G
Methyl-4-hydroxy-3-(trifluoromethyl)benzoate 98+%
Methyl-4-hydroxy-3-(trifluoromethyl)benzoate
Formel: C₉H₇F₃O₃
Molecular Weight: 220,1500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06204091
CAS-Nummer: 115933-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR