3-Fluoroprop-1-yl trifluoromethanesulphonate 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Fluoroprop-1-yl triflate

PC2905-5G
APOSPC2905-5GEA 596 EUR
APOSPC2905-5G
3-Fluoroprop-1-yl trifluoromethanesulphonate 95%
3-Fluoroprop-1-yl trifluoromethanesulphonate
Formel: C₄H₆F₄O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD07784315
CAS-Nummer: 180597-96-0
UN: 2920
ADR: 8+3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR