2H,2H,3H,3H-Perfluorooctanoic acid
Lieferant: Apollo Scientific
|
Gefahr
|
PC2908-1G
APOSPC2908-1GEA
188
EUR
APOSPC2908-1G
2H,2H,3H,3H-Perfluorooctanoic acid
2H,2H,3H,3H-Perfluorooctanoic acid
Formel:
C₈H₅F₁₁O₂ Molecular Weight: 342,107 g/mol Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08460490 CAS-Nummer: 914637-49-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...