1,1,1-Trifluoropropan-2-amine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,1,1-Trifluoropropan-2-amine hydrochloride, 2-Amino-1,1,1-trifluoropropane hydrochloride

PC2929-1G
APOSPC2929-1GEA 21.9 EUR
APOSPC2929-1G
1,1,1-Trifluoropropan-2-amine hydrochloride
1-(Trifluoromethyl)ethylamine hydrochloride
Formel: C₃H₆F₃N·ClH
Molecular Weight: 149.543 g/mol
Schmelzpunkt: 218…221 °C (Subl.)
Lagertemperatur: Raumtemperatur
MDL: MFCD04972986
CAS-Nummer: 2968-32-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR