Ethyl-4-hydroxy-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

PC2935-1G
APOSPC2935-1GEA 149 EUR
APOSPC2935-1G
Ethyl-4-hydroxy-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate
Ethyl-4-hydroxy-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate
Formel: C₁₃H₁₀F₃NO₃S
Molecular Weight: 317,29 g/mol
Schmelzpunkt: 121…123 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00816788
CAS-Nummer: 227199-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR