1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)-3-methylpiperazin

Lieferant: Apollo Scientific

PC2946-1G
APOSPC2946-1GEA 288 EUR
APOSPC2946-1G
1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)-3-methylpiperazin
1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)-3-methylpiperazin
Formel: C₁₁H₁₃ClF₃N₃
Schmelzpunkt: 63…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692451
CAS-Nummer: 914637-57-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR