1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperidine-4-carboxamide
Lieferant: Apollo Scientific
PC2949-1G
APOSPC2949-1GEA
242
EUR
APOSPC2949-1G
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperidine-4-carboxamide
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperidine-4-carboxamide
Formel:
C₁₂H₁₃ClF₃N₃O Schmelzpunkt: 187…188 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00172305 CAS-Nummer: 338780-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...