1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-piperidinecarboxylic acid
Lieferant: Apollo Scientific
PC2951-1G
APOSPC2951-1GEA
0
EUR
APOSPC2951-1G
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-piperidinecarboxylic acid
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-piperidinecarboxylic acid
Formel:
C₁₂H₁₂ClF₃N₂O₂ Schmelzpunkt: 86…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05664789 CAS-Nummer: 874800-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...