1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-piperidine-4-ol

Lieferant: Apollo Scientific

PC2952-1G
APOSPC2952-1GEA 376 EUR
APOSPC2952-1G
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-piperidine-4-ol
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-piperidine-4-ol
Formel: C₁₁H₁₂ClF₃N₂O
Molecular Weight: 280,68 g/mol
Schmelzpunkt: 74…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00202495
CAS-Nummer: 306976-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR