1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]pyrrolidine-2-carboxylic acid

Lieferant: Apollo Scientific

PC2953-1G
APOSPC2953-1GEA 434 EUR
APOSPC2953-1G
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]pyrrolidine-2-carboxylic acid
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]pyrrolidine-2-carboxylic acid
Formel: C₁₁H₁₀ClF₃N₂O₂
Schmelzpunkt: 114…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07657733
CAS-Nummer: 1188435-80-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR