5-Oxo-1-[4-(trifluoromethyl)phenyl]pyrrolidine-3-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

PC2954-250MG
APOSPC2954-250MGEA 134 EUR
APOSPC2954-250MG
5-Oxo-1-[4-(trifluoromethyl)phenyl]pyrrolidine-3-carboxylic acid 95%
5-Oxo-1-[4-(trifluoromethyl)phenyl]pyrrolidine-3-carboxylic acid
Formel: C₁₂H₁₀F₃NO₃
Molecular Weight: 273.21 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00665831
CAS-Nummer: 2357-27-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR