2,3-Dihydro-6-(trifluoromethyl)benzo[1,4]thiazin-3-one 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC2954E-100MG
PC2954E-500MG
APOSPC2954E-100MGEA
0
EUR
APOSPC2954E-100MG
APOSPC2954E-500MG
2,3-Dihydro-6-(trifluoromethyl)benzo[1,4]thiazin-3-one 97%
2,3-Dihydro-6-(trifluoromethyl)benzo[1,4]thiazin-3-one
Formel:
C₉H₆F₃NOS Schmelzpunkt: 192…194 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00178754 CAS-Nummer: 716-82-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...