3-Fluor-4-nitroanisol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-4-methoxy-1-nitrobenzene
PC2959-25G
PC2959-5G
PC2959-100G
APOSPC2959-25GEA
55.1
EUR
APOSPC2959-25G
APOSPC2959-5G
APOSPC2959-100G
3-Fluor-4-nitroanisol 98%
3-Fluor-4-nitroanisol
Formel:
C₇H₆FNO₃ Schmelzpunkt: 56…58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115632 CAS-Nummer: 446-38-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...