1,2-Diiodtetrafluorbenzol 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC2969-1G
PC2969-25G
PC2969-5G
APOSPC2969-1GEA
36.4
EUR
APOSPC2969-1G
APOSPC2969-25G
APOSPC2969-5G
1,2-Diiodtetrafluorbenzol 97%
1,2-Diiodtetrafluorbenzol
Formel:
C₆F₄I₂ Schmelzpunkt: 49…50 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00019011 CAS-Nummer: 2708-97-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...