2-Chlor-4-(trifluormethyl)benzolsulfonamid

Lieferant: Apollo Scientific

PC2976-1G
APOSPC2976-1GEA 79.9 EUR
APOSPC2976-1G
2-Chlor-4-(trifluormethyl)benzolsulfonamid
2-Chlor-4-(trifluormethyl)benzolsulfonamid
Formel: C₇H₅ClF₃NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00179384
CAS-Nummer: 146533-47-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR