2-(Trifluormethyl)benzolsulfonamid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Trifluormethyl)benzolsulfonsäureamid, α,α,α-Trifluor-o-toluolsulfonamid
PC2977-1G
PC2977-25G
PC2977-5G
APOSPC2977-1GEA
21.9
EUR
APOSPC2977-1G
APOSPC2977-25G
APOSPC2977-5G
2-(Trifluormethyl)benzolsulfonamid
2-(Trifluormethyl)benzolsulfonamid
Formel:
C₇H₆F₃NO₂S Schmelzpunkt: 180…184 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042420 CAS-Nummer: 1869-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...