5-Fluor-2-(trifluormethyl)anilin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
α,α,α,5-Tetrafluoro-o-toluidine
PC2986-5G
PC2986-10G
PC2986-1G
APOSPC2986-5GEA
81.2
EUR
APOSPC2986-5G
APOSPC2986-10G
APOSPC2986-1G
5-Fluor-2-(trifluormethyl)anilin 98%
5-Fluor-2-(trifluormethyl)anilin
Formel:
C₇H₅F₄N Molecular Weight: 179.12 g/mol Schmelzpunkt: 29…32 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660353 CAS-Nummer: 827-20-3 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...