2-Fluoro-4-nitropyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC2992-250MG
APOSPC2992-250MGEA
46.4
EUR
APOSPC2992-250MG
2-Fluoro-4-nitropyridine 95%
2-Fluoro-4-nitropyridine
Formel:
C₅H₃FN₂O₂ Schmelzpunkt: 46 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04112489 CAS-Nummer: 18614-46-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...