3-Carboxy-4-fluorphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Carboxy-4-fluorophenylboronic Acid, 3-Carboxy-4-fluorobenzeneboronic acid
PC2998-25G
PC2998-5G
PC2998-100G
APOSPC2998-25GEA
89.9
EUR
APOSPC2998-25G
APOSPC2998-5G
APOSPC2998-100G
3-Carboxy-4-fluorphenylboronsäure
3-Carboxy-4-fluorphenylboronsäure
Formel:
C₇H₆BFO₄ Schmelzpunkt: 217…219 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095129 CAS-Nummer: 872460-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...