4-(Ethoxycarbonyl)-3-fluorphenylboronsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Ethoxycarbonyl-3-fluorobenzeneboronic acid, 4-Ethoxycarbonyl-3-fluorophenylboronic acid

PC2999-1G
APOSPC2999-1GEA 46.4 EUR
APOSPC2999-1G
4-(Ethoxycarbonyl)-3-fluorphenylboronsäure 95%
4-(Ethoxycarbonyl)-3-fluorphenylboronsäure
Formel: C₉H₁₀BFO₄
Molecular Weight: 211,98 g/mol
Schmelzpunkt: 158…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06656274
CAS-Nummer: 874288-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR