6-(Benzylamino)-2-chloro-5-fluoronicotinic acid

Lieferant: Apollo Scientific

PC300004-1G
APOSPC300004-1GEA 311 EUR
APOSPC300004-1G
6-(Benzylamino)-2-chloro-5-fluoronicotinic acid
6-(Benzylamino)-2-chloro-5-fluoronicotinic acid
Formel: C₁₃H₁₀ClFN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22126065
CAS-Nummer: 1417334-56-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR