3-(Trifluoromethyl)-1H-pyrazole-4-carbaldehyde

Lieferant: Apollo Scientific
Achtung

PC300005-1G
APOSPC300005-1GEA 0 EUR
APOSPC300005-1G
3-(Trifluoromethyl)-1H-pyrazole-4-carbaldehyde
3-(Trifluoromethyl)-1H-pyrazole-4-carbaldehyde
Formel: C5H3F3N2O
Lagertemperatur: Raumtemperatur
MDL: MFCD07375188
CAS-Nummer: 1001020-14-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR