3-(Trifluoromethoxy)benzeneacetyl chloride

Lieferant: Apollo Scientific
Gefahr

PC300007-50MG PC300007-250MG PC300007-500MG
APOSPC300007-50MGEA 123 EUR
APOSPC300007-50MG APOSPC300007-250MG APOSPC300007-500MG
3-(Trifluoromethoxy)benzeneacetyl chloride
3-(Trifluoromethoxy)benzeneacetyl chloride
Formel: C₉H₆ClF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD17018703
CAS-Nummer: 929612-79-3
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR