6-Bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine

Lieferant: Apollo Scientific
Achtung

PC300501-250MG
APOSPC300501-250MGEA 130 EUR
APOSPC300501-250MG
6-Bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine
6-Bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine
Formel: C₈H₄BrF₃N₂
Molecular Weight: 265,03 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD22577291
CAS-Nummer: 1417334-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR