6-(Trifluoromethyl)-2H-1,4-benzoxazin-3(4H)-one 95%

Lieferant: Apollo Scientific

Synonyme: 3,4-Dihydro-3-oxo-6-(trifluoromethyl)-2H-1,4-benzoxazine, 6-(Trifluoromethyl)-4H-benzo[b][1,4]oxazin-3-one

PC300507-1G
APOSPC300507-1GEA 203 EUR
APOSPC300507-1G
6-(Trifluoromethyl)-2H-1,4-benzoxazin-3(4H)-one 95%
6-(Trifluoromethyl)-2H-1,4-benzoxazin-3(4H)-one
Formel: C₉H₆F₃NO₂
Molecular Weight: 217,145 g/mol
Schmelzpunkt: 152…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02648641
CAS-Nummer: 189940-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR