2-Ethyl-5-(trifluoromethyl)-1,3-benzoxazole
Lieferant: Apollo Scientific
PC300511-1G
APOSPC300511-1GEA
143
EUR
APOSPC300511-1G
2-Ethyl-5-(trifluoromethyl)-1,3-benzoxazole
2-Ethyl-5-(trifluoromethyl)-1,3-benzoxazole
Formel:
C₁₀H₈F₃NO Molecular Weight: 215,172 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19375803 CAS-Nummer: 1267428-36-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...