2-(Chloromethyl)-5-(trifluoromethyl)-1,3-benzoxazole 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Chloromethyl)-5-(trifluoromethyl)benzo[d]oxazole

PC300514-1G
APOSPC300514-1GEA 263 EUR
APOSPC300514-1G
2-(Chloromethyl)-5-(trifluoromethyl)-1,3-benzoxazole 95%
2-(Chloromethyl)-5-(trifluoromethyl)-1,3-benzoxazole
Formel: C₉H₅ClF₃NO
Molecular Weight: 235,59 g/mol
Schmelzpunkt: 50…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD10696619
CAS-Nummer: 131337-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR